Learn More
Alfa Aesar™ C6 Ceramide
Manufacturer:  Alfa Aesar™ J67034LB0
 View more versions of this product
Catalog No. AAJ67034LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
-30°C to -10°C
C6 Ceramide
C24H47NO3
N-Hexanoyl-D-erythro-sphingosine;
White
Crystalline solid
124753-97-5
397.64