Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C6orf120 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546381
Description
Peptide sequence: LLLLASQVLS PGSCADEEEV PEEWVLLHVV QGQIGAGNYS YLRLNHEGKI.
C6orf120 may be involved in induction of apoptosis in CD4(+) T- cells, but not CD8(+) T-cells or hepatocytes.
Specifications
C6orf120 | |
Polyclonal | |
Unconjugated | |
C6orf120 | |
bA160E12.4; C6orf120; chromosome 6 open reading frame 120; UPF0669 protein C6orf120 | |
Rabbit | |
Affinity chromatography | |
RUO | |
387263 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7Z4R8 | |
C6orf120 | |
Synthetic peptide directed towards the N-terminal of human C6orf120 (aa 16-65). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction