Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTCHD4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548192

Catalog No. PA548192

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Specifications

Specifications

PTCHD4
Polyclonal
PBS with 0.09% sodium azide
Q6ZW05
Patched domain-containing protein 4, PTCHD4, C6orf138
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
442213
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PTCHD4
Liquid
PTCHD4
KLH conjugated synthetic peptide between 563-591 amino acids from the Central region of human C6orf138.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit