Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

C6orf146 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19146220UL

 View more versions of this product

Catalog No. NBP19146220



Description

Description

C6orf146 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

C6orf146
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
FAM217A
Synthetic peptide directed towards the middle region of human C6orf146. Peptide sequence ETLPAPNWNLKHGNSSVEENFTDESDLSENEKTNDTLLSYFKKVDLNLKP.
Immunogen affinity purified
RUO
Primary
222826
Western Blot
Western Blot 1:1000
NP_775834
C6orf146, chromosome 6 open reading frame 146, family with sequence similarity 217, member A, hypothetical protein LOC222826
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit