Learn More
C6orf182 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 249571AP
Catalog No. 50-173-4643
Add to cart
Description
Description
C6orf182 also named as Centrosomal protein of 57 kDa related protein is a 460 amino-acid protein, which belongs to translokin family. C6orf182 as centrosomal protein may be required for microtubule attachment to centrosomes and localizes in the cytoplasm. The function of C6orf182 need to be further studied.
Specifications
Specifications
C6orf182
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IYX8, Q8VDS7
C6orf182, Centrosomal protein 57kDa-like protein 1, Cep57 related protein, CEP57L1, Cep57R
Rabbit
IgG
150 μL
-20°C
Primary
103268, 285753
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CEP57L1
Liquid
CEP57L1
C6orf182 Fusion Protein Ag21054
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate