Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf211 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548750

Catalog No. PA548750

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with rat based on sequence homology.

O-methyltransferase that methylates glutamate residues of target proteins to form gamma-glutamyl methyl ester residues. Methylates PCNA, suggesting it is involved in the DNA damage response.
Specifications

Specifications

C6orf211
Polyclonal
PBS with 0.09% sodium azide
Q9H993
UPF0364 protein C6orf211, C6orf211
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79624
Western Blot
Unconjugated
ARMT1
Liquid
ARMT1
KLH conjugated synthetic peptide between 29-57 amino acids from the N-terminal region of human C6orf211.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit