Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C7 Polyclonal Antibody
C7 Polyclonal Antibody
Supplier: Thermo Scientific PA568370
Description
C7 Polyclonal Antibody for Western Blot

Specifications
C7 | |
Polyclonal | |
Unconjugated | |
C7 | |
A530026G17; C7; complement component 7; complement component C7; Gm1238 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
109828, 730 | |
-20°C | |
Liquid |
Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P10643 | |
C7 | |
Synthetic protein corresponding to residues near the C terminal of human C7. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction