Learn More
Aviva Systems Biology Corp C7ORF29 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP52601_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C7orf29. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C7orf29. Alternate names: C7orf29, ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60779
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.