Learn More
C7orf30 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 228381AP
Catalog No. 50-173-3952
Add to cart
Specifications
Specifications
C7orf30
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96EH3
C7orf30, MALSU1
Rabbit
IgG
150 μL
-20°C
Primary
115416
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MALSU1
Liquid
MALSU1
C7orf30 Fusion Protein Ag18286
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human