Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C7orf31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113207
Description
C7orf31 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
C7orf31 is a protein coding gene.
Specifications
C7orf31 | |
Polyclonal | |
Unconjugated | |
C7orf31 | |
C7orf31; chromosome 7 open reading frame 31; uncharacterized protein C7orf31 | |
Rabbit | |
Protein G | |
RUO | |
136895 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8N865 | |
C7orf31 | |
Recombinant Human Uncharacterized protein C7orf31 protein (163-256AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction