Learn More
Abnova™ C7orf38 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00221786T01
Catalog No. 89-937-618
Add to cart
Description
Description
Specifications
Specifications
Western Blot
C7orf38 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 63.14kDa
Transient overexpression cell lysate was tested with anti-C7orf38 antibody (Mfr. No. H00221786-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_145111.2
pCMV-C7orf38 full-length
100 μL
Human