Learn More
Aviva Systems Biology Corp C7orf42 Antibody - N-terminal region (ARP47385_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47385_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C7orf42. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C7orf42. Alternate names: FLJ10099, FLJ13090, C7orf42ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59534