Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C8orf46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571004
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Required for neurogenesis in the neural plate and retina. Strongly cooperates with neural bHLH factors to promote neurogenesis.
Specifications
C8orf46 | |
Polyclonal | |
Unconjugated | |
C8orf46 | |
C8orf46; chromosome 8 open reading frame 46; uncharacterized protein C8orf46; Vexin; VXN | |
Rabbit | |
Affinity chromatography | |
RUO | |
254778 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TAG6 | |
C8orf46 | |
synthetic peptide directed towards the C-terminal region of human C8orf46. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction