Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ C9orf103 E. coli Recombinant Protein, TMW: 23.4kDa

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Supplier:  Novus Biologicals™ NBP2522390.1MG

 View more versions of this product

Catalog No. NBP25223901


Add to cart

Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-187 of E.coli C9orf103 The Recombinant E. coli C9orf103 Protein is derived from E. coli. The Recombinant E. coli C9orf103 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

1mg/mL
20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 10% glycerol, 1mM DTT
TMW: 23.4kDa
0.1mg
bA522I20.2, C9orf103, chromosome 9 open reading frame 103, EC 2.7.1.12, glucokinase-like protein, Gluconate kinase, gluconokinase-like protein, idnK, gluconokinase homolog (E. coli), probable gluconokinase
Specific activity is > 80 units/mg, which one unit will convert 1.0 umole of D-gluconate to 6-phospho-D-gluconate per minute at pH 8.0 at 37C.
Yes
SDS-Page
414328
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
C9orf103
Unlabeled
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only and is not approved for use in humans or in clinical diagnosis