Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ C9orf103 E. coli Recombinant Protein, TMW: 23.4kDa
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
$206.00 - $436.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 10% glycerol, 1mM DTT |
Gene ID (Entrez) | 414328 |
Molecular Weight | TMW: 23.4kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2522392
|
Novus Biologicals™
NBP2522390.02MG |
0.02mg |
Each for $206.00
|
|
NBP25223901
|
Novus Biologicals™
NBP2522390.1MG |
0.1mg |
Each for $436.00
|
|
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-187 of E.coli C9orf103 The Recombinant E. coli C9orf103 Protein is derived from E. coli. The Recombinant E. coli C9orf103 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
1mg/mL | |
20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 10% glycerol, 1mM DTT | |
TMW: 23.4kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
C9orf103 | |
Unlabeled |
SDS-Page | |
414328 | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
bA522I20.2, C9orf103, chromosome 9 open reading frame 103, EC 2.7.1.12, glucokinase-like protein, Gluconate kinase, gluconokinase-like protein, idnK, gluconokinase homolog (E. coli), probable gluconokinase | |
Specific activity is > 80 units/mg, which one unit will convert 1.0 umole of D-gluconate to 6-phospho-D-gluconate per minute at pH 8.0 at 37C. | |
Yes |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title