Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C9orf114 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140198
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: CGPGEHGQRI EWRKWKQQKK EEKKKWKDLK LMKKLERQRA QEEQAKRLEE EEAAAEKEDR GRPYTLS.
C9orf114 is a protein coding gene. Required for association of the centrosomes with the poles of the bipolar mitotic spindle during metaphase. Also involved in chromosome alignment. May promote centrosome maturation probably by recruiting A-kinase anchor protein AKAP9 to centrosomes in early mitosis. Binds specifically to miRNA MIR145 hairpin, regulates MIR145 expression at a postranscriptional level.
Specifications
C9orf114 | |
Polyclonal | |
Unconjugated | |
SPOUT1 | |
9930028P05; AL033358; C9orf114; CENP-32; Centromere protein 32; D2Wsu81e; DNA segment, Chr 2, Wayne State University 81, expressed; HSPC109; Kinetochore-associated protein; LOC499770; Putative methyltransferase C9orf114; putative methyltransferase C9orf114 homolog; similar to LOC495800 protein; SPOUT domain containing methyltransferase 1; SPOUT domain-containing methyltransferase 1; Spout1; uncharacterized protein C9orf114 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51490 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T280 | |
SPOUT1 | |
Recombinant protein corresponding to Human SPOUT1. Recombinant protein control fragment (Product #RP-109602). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction