Learn More
Aviva Systems Biology Corp CFAP157 Antibody - N-terminal region (ARP54415_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54415_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf117. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human C9orf117. Alternate names: RP11-56D16.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61201