Learn More
Aviva Systems Biology Corp C9orf153 Antibody - middle region (ARP54403_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54403_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf153. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C9orf153. Alternate names: bA507D14.1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61197