Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf156 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549602

Catalog No. PA549602

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse and rat based on sequence homology.

Hydrolyzes acyl-CoA thioesters (in vitro). Has a preference for substrates with medium chain length (C10-C14). Inactive towards substrates with C18 or C20 aliphatic chains. Its physiological substrate is not known.
Specifications

Specifications

C9orf156
Polyclonal
PBS with 0.09% sodium azide
Q9BU70
Nef-associated protein 1, 312-, Thioesterase NAP1, C9orf156, NAP1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51531
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TRMO
Liquid
TRMO
KLH conjugated synthetic peptide between 365-391 amino acids from the C-terminal region of human C9orf156.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit