Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen C9orf4
Immunogen Synthetic peptides corresponding to C9ORF4 The peptide sequence was selected from the N terminal of C9ORF4. Peptide sequence PAACAASPADDGAGPGGRGPRGRARGDTGADEAVPRHDSSYGTFAGEFYD.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1599620 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15996120UL
20ul Each for $149.00
Add to cart
 
NBP159961 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159961
100 ul Each for $329.00
Add to cart
 
Description

Description

C9orf4 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

C9orf4
Rabbit
Polyclonal
Unconjugated
23732
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to C9ORF4 The peptide sequence was selected from the N terminal of C9ORF4. Peptide sequence PAACAASPADDGAGPGGRGPRGRARGDTGADEAVPRHDSSYGTFAGEFYD.
Primary
IgG
Brain protein CG-6, C9orf4, CG6, CG-6, chromosome 9 open reading frame 4, ferric-chelate reductase 1-like, hypothetical protein LOC23732
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit