Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C9orf64 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563038
Description
Immunogen sequence: QDEHKCVVRY RGKTYSGYWS LCAAVNRALD EGIPITSASY YATVTLDQVR NILRSDTDVS MPLVEERHRI LNETGKILLE KFGGS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 82%.
Involved in salvaging queuosine.
Specifications
C9orf64 | |
Polyclonal | |
Unconjugated | |
C9orf64 | |
C9orf64; chromosome 9 open reading frame 64; Queuosine salvage protein; UPF0553 protein; UPF0553 protein C9orf64 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84267 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T6V5 | |
C9orf64 | |
Recombinant protein corresponding to Human C9orf64. Recombinant protein control fragment (Product #RP-93035). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction