Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf72 Mouse anti-Human, Mouse, Rat, Clone: GT779, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527865

Catalog No. PIMA527865


Add to cart

Description

Description

Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse brain, rat brain Predicted Reactivity: Xenopus laevis (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis
Specifications

Specifications

C9orf72
Monoclonal
1.0 mg/mL
PBS with no preservative
Q96LT7
ALSFTD, FTDALS, chromosome 9 open reading frame 72
Mouse
IgG1
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
203228, 313155, 73205
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
GT779
Unconjugated
C9orf72
Liquid
3110043O21Rik, C9orf72
Human C9orf72
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit