Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf72 peptide 5 Rat anti-Human, Clone: 5F6, MilliporeSigma™

Rat Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen C9orf72 peptide 5
Clone 5F6
Concentration 1 mg/mL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN2310100
View Documents
MilliporeSigma
MABN2310100UG
100 μg
Each for $387.00
Add to cart
 
MABN231025
View Documents
MilliporeSigma
MABN231025UG
25 μg
Each for $153.40
Add to cart
 
Description

Description

C9orf72 peptide 5 Monoclonal antibody specifically detects C9orf72 peptide 5 in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

C9orf72 peptide 5
1 mg/mL
Unconjugated
Rat
Human
Ovalbumin-conjugated linear peptide corresponding to 20 amino acids from the N-terminal half (peptide 5) of human C9orf72.
Monoclonal
Protein G purified
5F6
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
RUO
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit