Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C9orf72 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109471
Description
Immunogen sequence: SPAVAKTEIA LSGKSPLLAA TFAYWDNILG PRVRHIWAPK TEQVLLSDGE ITFLANHTLN GEILRNAESG AIDVKFFVLS EKGVIIVSLI FDGNWNGDRS TYGLSIILPQ TELSFYLPLH RVCVDRLTHI IRKGRIWMHK ERQENVQKII LEGTERMEDQ GQSIIPMLTG EVIPVMELLS SMKSHSVPEE ID.
C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.
Specifications
C9orf72 | |
Polyclonal | |
Unconjugated | |
C9orf72 | |
3110043O21Rik; AI840585; ALSFTD; C9orf72; chromosome 9 open reading frame 72; FTDALS; FTDALS1; Guanine nucleotide exchange C9orf72; Guanine nucleotide exchange C9orf72 homolog; protein C9orf72; protein C9orf72 homolog; RGD1359108; RIKEN cDNA 3110043O21 gene; similar to RIKEN cDNA 3110043O21 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
203228, 313155, 73205 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q66HC3, Q96LT7 | |
C9orf72, RGD1359108 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 9-200 of human C9orf72 (NP_0012429831). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction