Learn More
Aviva Systems Biology Corp C9orf75 Antibody - middle region (ARP55692_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55692_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C9orf75. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human C9orf75. Alternate names: FLJ90254, MGC131933, RP11-350O14.7, DFNB79, C9orf75ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61582