Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C9orf96 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS15350R
Description
STKLD1 (Serine/Threonine Kinase Like Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includetransferase activity, transferring phosphorus-containing groupsandprotein tyrosine kinase activity.Specifications
C9orf96 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
STKLD1 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
169436 |
STKLD1 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8NE28 | |
C9orf96; LOC169436; probable inactive protein kinase-like protein SgK071; RP11-244N20.8; serine/threonine kinase like domain containing 1; serine/threonine kinase-like domain containing 1; Serine/threonine kinase-like domain-containing protein 1; Serine/threonine kinase-like domain-containing protein STKLD1; SGK071; Sk521; STKLD1; Sugen kinase 071 | |
KLH conjugated synthetic peptide derived from human C9orf96. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title