Learn More
Selleck Chemical LLCSupplier Diversity Partner CA-4948 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S85622MG
CA-4948 is a reversible kinase inhibitor that modulates IRAK4 function in both the toll-like receptor (TLR) and interleukin 1 receptor (IL-1R) signaling cascades, and demonstrates pharmacodynamic and antitumor activity in in vitro and in vivo nonclinical models.
Catalog No. 50-194-8127