Learn More
Sterlitech CorporationSupplier Diversity Partner Cellulose Acetate Syringe Filters, 0.22 Micron, 25mm, 100/Pk DFS Item
Manufacturer:  Sterlitech CorporationSupplier Diversity Partner 1470365
Cellulose Acetate Syringe Filter Applications:* Sterilize biological fluids, serum or media additives* Sample preparations of aqueous solutions* Sample preparation of protein-based HPLC solutions* High throughput, low binding filter units for non-sterile aqueous filtrations* Filtrations of tissue culture media* High throughput for sterile or non-sterile clarification of even the most viscous proteinaceous Solutions* Filter probe and hybridization solutions to reduce backgrounds
Catalog No. 50-195-2316