Learn More
Abnova™ CA11 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00000770T02
Catalog No. 89-937-722
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CA11 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 36.19kDa
Transient overexpression cell lysate was tested with anti-CA11 antibody (Mfr. No. H00000770-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_001217.3
pCMV-CA11 full-length
100 μL
Human