Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA19-9/Sialyl Lewis A Rabbit anti-Human, Allophycocyanin/Cy7, Clone: CA19.9/1390R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254585APCCY7

Catalog No. N254585APC7


Add to cart

Description

Description

CA19-9/Sialyl Lewis A Monoclonal antibody specifically detects CA19-9/Sialyl Lewis A in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CA19-9/Sialyl Lewis A
CA19.9/1390R
Flow Cytometry
Conjugated, Purified
Purified human CA19-9 protein
Protein A purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
Rabbit
IgG Kappa
0.1 mL
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit