Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ CA19-9/Sialyl Lewisa (GI Tumor Marker) Mouse Monoclonal Antibody with BSA and azide
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CA19-9/Sialyl Lewisa (GI Tumor Marker), Mouse Monoclonal Antibody.
Supplier: Epredia™ MS9085R1
Description
Mucin glucoprotein is a sialyl Lewisa structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewisa and/or Lewisb blood group antigens. In normal tissues, sialyl Lewisa antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas.Host Species: Mouse
Clone: SPM110
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Immunogen: Purified human CA19-9
Molecular Weight: >400kDa
Positive Control: Colon CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) | |
Monoclonal | |
Unconjugated | |
Purified human CA19-9 | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
SPM110 | |
Mouse | |
1 mL | |
Oncogenesis | |
Human | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction