Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CA19-9/Sialyl Lewis a (GI Tumor Marker), Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CA19-9/Sialyl Lewisa (GI Tumor Marker), Mouse Monoclonal Antibody.

Specifications

Antigen CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker)
Clone SPM110
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS9085R1
View Documents
Thermo Scientific™
MS9085R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CA19-9/Sialyl Lewisa (GI Tumor Marker), Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Mucin glucoprotein is a sialyl Lewisa structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewisa and/or Lewisb blood group antigens. In normal tissues, sialyl Lewisa antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas.

Host Species: Mouse

Clone: SPM110

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Immunogen: Purified human CA19-9

Molecular Weight: >400kDa

Positive Control: Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker)
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG1, κ
SPM110
Monoclonal
Murine
Oncogenesis
Purified human CA19-9
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit