Learn More
CA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 169611AP
Catalog No. 50-173-1296
Add to cart
Description
Description
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
Specifications
Specifications
CA2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00918, P00920, P27139
CA II, CA2, CAC, CAII, Car2, Carbonate dehydratase II, Carbonic anhydrase 2, Carbonic anhydrase C, carbonic anhydrase II
Rabbit
IgG
150 μL
-20°C
Primary
12349, 54231, 760
Immunoprecipitation, Western Blot
Unconjugated
CA2
Liquid
Ca2
CA2 Fusion Protein Ag10680
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat