Learn More
Abnova™ CA3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000761T02
Catalog No. 89-937-736
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CA3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 29.6kDa
Transient overexpression cell lysate was tested with anti-CA3 antibody (Mfr. No. H00000761-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_005181
pCMV-CA3 full-length
100 μL
Human