Learn More
CA7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 136701AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-172-8924
Add to cart
Description
Description
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known; however, it may have a role in inherited renal abnormalities of bicarbonate transport.
Specifications
Specifications
CA7
Polyclonal
Unconjugated
CA7
CA VII, CA7, Carbonate dehydratase VII, Carbonic anhydrase 7, carbonic anhydrase VII, CAVII
Rabbit
Antigen Affinity Chromatography
RUO
12354, 291819, 766
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P43166, Q9ERQ8
Ca7, Car7
CA7 Fusion Protein Ag4649
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG