Learn More
CAB39L Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 161371AP
Catalog No. 50-173-0746
Add to cart
Description
Description
The function of this protein remains unknown.
Specifications
Specifications
CAB39L
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H9S4
Antigen MLAA 34, CAB39L, MO25 BETA, Mo25 like protein, MO2L, RP11 103J18.3
Rabbit
IgG
150 μL
-20°C
Primary
81617
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CAB39L
Liquid
CAB39L
CAB39L Fusion Protein Ag9181
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human