Learn More
Abnova™ CABC1 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00056997T02
Catalog No. 89-937-763
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CABC1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 71.9kDa
Transient overexpression cell lysate was tested with anti-CABC1 antibody (Mfr. No. H00056997-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_020247.4
pCMV-CABC1 full-length
100 μL
Human