Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Cabinet Labels: SAFETY EQUIPMENT

Clearly identify location of chemicals, safety equipment, and other supplies in cabinets and storage areas. Brady™ Cabinet Labels are designed for any size cabinet, including single door cabinets and low capacity cabinets.

Manufacturer:  Brady™ 60302

 View more versions of this product

Catalog No. 19-104-788


  Check Availability
Add to cart

Specifications

Description

  • Clearly identify cabinets containing chemicals and safety equipment
  • Specially designed for placement on any size cabinet, including onedoor and low capacity cabinets
  • Made of B-302 self-adhesive laminated polyester
Description & Specifications

Specifications

Acrylic,Cold temperature,Permanent
SAFETY EQUIPMENT
Rectangle
0.01 in.
Cabinet Label
Green,White
Laminated
W x H: 12 x 3.5 in.
Self Sticking Polyester (B-302)
1/Pk.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit