Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CABLES2 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS5744R
Description
Unknown. Probably involved in G1-S cell cycle transition.Specifications
CABLES2 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CABLES2 | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
81928 |
CABLES2 | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9BTV7 | |
C20orf150; CABLES2; Cdk5 and Abl enzyme substrate 2; CDK5 and ABL1 enzyme substrate 2; dJ908M14.2; Ik3-2; Interactor with CDK3 2 | |
KLH conjugated synthetic peptide derived from human CABLES2. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title