Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CABP2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549161
Description
This gene belongs to a subfamily of calcium binding proteins that share similarity to calmodulin. Like calmodulin, these family members can likely stimulate calmodulin-dependent kinase II and the protein phosphatase calcineurin. Calcium binding proteins are an important component of calcium mediated cellular signal transduction. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jan 2016]
Specifications
CABP2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9NPB3 | |
CABP2 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CABP2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CABP2 | |
Calcium-binding protein 2, CaBP2, CABP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51475 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction