Learn More
Invitrogen™ CABP7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589372
Description
Immunogen sequence: PKLSTSGIPE KFHGTDFDTV FWKCDMQKLT VDELKRLLYD TFCEHLSMKD IENIIMTEEE SHLGTAEECP VDVETCSNQQ I; Positive Samples: Mouse brain, Mouse lung, Mouse kidney, Rat lung, Rat kidney; Cellular Location: Cell membrane, Cytoplasm, Golgi apparatus, Single-pass type IV membrane protein, perinuclear region, trans-Golgi network membrane.
Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP7 (Calcium-binding protein 7), contains two EF-hand domains for calcium binding and shares 70% homology with CaBP8 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. CaBP7 and 8 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
Specifications
CABP7 | |
Polyclonal | |
Unconjugated | |
CABP7 | |
CABP7; calcium binding protein 7; calcium-binding protein 7; calcium-binding protein CaBP7; CALN2; calneuron 2; calneuron II; Calneuron-2; calneuron-II; rCaBP7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
164633, 192650, 360970 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q66H96, Q86V35, Q91ZM8 | |
CABP7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 100-180 of human CABP7 (NP_8723331). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.