Learn More
CABP7 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 176751AP
Catalog No. 50-173-1716
Add to cart
Description
Description
Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP7 (Calcium-binding protein 7), contains two EF-hand domains for calcium binding and shares 70% homology with CaBP8 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. CaBP7 and 8 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
Specifications
Specifications
CABP7
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86V35
CABP7, calcium binding protein 7, CALN2, Calneuron 2, Calneuron II
Rabbit
IgG
150 μL
-20°C
Primary
164633
Western Blot
Unconjugated
CABP7
Liquid
CABP7
CABP7 Fusion Protein Ag11654
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human