Learn More
CACNA1G Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: S178A-9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259322UV
Catalog No. NBP259322UV
Add to cart
Description
Description
CACNA1G Monoclonal antibody specifically detects CACNA1G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
CACNA1G
S178A-9
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
CACNA1G
Fusion protein amino acids 2052-2172 (cytoplasmic C-terminus) of mouse Cav3.1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects <200kDa. Does not cross-react with Cav3.2.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit
Mouse
IgG1
100 ul
Primary
8913
Human, Mouse, Rat