Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNA2D2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310676100UL
Description
CACNA2D2 Polyclonal specifically detects CACNA2D2 in Human samples. It is validated for Western Blot.Specifications
CACNA2D2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
alpha 2 delta calcium channel subunit, CACNA2D, calcium channel, voltage-dependent, alpha 2/delta subunit 2, gene 26, KIAA0558Voltage-gated calcium channel subunit alpha-2/delta-2, LUAC11.1, voltage-dependent calcium channel subunit alpha-2/delta-2 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human CACNA2D2 (NP_006021). Peptide sequence NCSRLFHAQRLTNTNLLFVVAEKPLCSQCEAGRLLQKETHSDGPEQCELV | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
9254 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction