Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB1 Mouse anti-Human, Mouse, Rat, Clone: S7-18, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527714

Catalog No. PIMA527714


Add to cart

Description

Description

FAM3C is a 227 amino acid secretory protein that belongs to a novel cytokine-like family with sequence similarity to FAM3 family. FAM3C is a signaling cytokine with a GG domain, hydrophobic leader sequence and an N-terminal signal peptide. FAM3C has a cytokine activity and shows a change in expression in pancreatic cancer-derived cells. It is ubiquitously expressed in most of the tissues. Reports suggest that FAM3C plays an important role in inner ear development and may thus be involved in cell differentiation and proliferation during inner ear embryogenesis.
Specifications

Specifications

CACNB1
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
P54283, Q02641, Q8R3Z5
CACNB1, CAB1, CACNLB1, voltage dependent L type calcium channel beta 1 subunit
Mouse
IgG1
100 μg
-20°C
Primary
12295, 50688, 782
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
S7-18
Unconjugated
CACNB1
Liquid
CACNB1
Synthetic peptide amino acids 19-34 of rat CavB1
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit