Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB2 Mouse anti-Human, Mouse, Rat, Clone: S8B-1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527713

Catalog No. PIMA527713


Add to cart

Description

Description

The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
Specifications

Specifications

CACNB2
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Q08289, Q8CC27, Q8VGC3
Cacnlb2, cacnb2, Voltage-dependent L-type calcium channel subunit beta-2, CAB2
Mouse
IgG1
100 μg
-20°C
Primary
116600, 12296, 783
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
S8B-1
Unconjugated
CACNB2
Liquid
Cacnb2
Synthetic peptide amino acids 189-205 of rat CavBeta2
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit