Learn More
CACNB2 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: S8b-1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP147607IR
Catalog No. NBP147607IR
Add to cart
Description
Description
CACNB2 Monoclonal antibody specifically detects CACNB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
Specifications
CACNB2
S8b-1
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q8VGC3
CAB2, CACNLB2, Calcium channel voltage-dependent subunit beta 2, calcium channel, voltage-dependent, beta 2 subunit, CAVB2, FLJ23743, Lambert-Eaton myasthenic syndrome antigen B, myasthenic (Lambert-Eaton) syndrome antigen B, MYSB, voltage-dependent L-type calcium channel subunit beta-2
Mouse
IgG1
0.1 ml
Primary
783
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Purified
CACNB2
Synthetic peptide amino acids 189-205 of rat CavBeta2
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 78 kDa. No cross reactivity against CavB1, CavB3, CavB4.