Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNG1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen CACNG1
Immunogen Synthetic peptides corresponding to Cacng1 (calcium channel, voltage-dependent, gamma subunit 1) The peptide sequence was selected from the middle region of Cacng1. Peptide sequence KNCSYFRHFNPGESSEIFEFTTQKEYSISAAAIAIFSLGFIIIGSICAFL.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16907620 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16907620UL
20ul Each for $149.00
Add to cart
 
NBP169076 View Documents Novus BiologicalsSupplier Diversity Partner
NBP169076
100 ul Each for $329.00
Add to cart
 
Description

Description

CACNG1 Polyclonal antibody specifically detects Antigen in Rat samples. It is validated for Western Blotting.
Specifications

Specifications

CACNG1
Rabbit
Polyclonal
Unconjugated
786
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to Cacng1 (calcium channel, voltage-dependent, gamma subunit 1) The peptide sequence was selected from the middle region of Cacng1. Peptide sequence KNCSYFRHFNPGESSEIFEFTTQKEYSISAAAIAIFSLGFIIIGSICAFL.
Primary
IgG
CACNLGDihydropyridine-sensitive L-type, skeletal muscle calcium channel subunit gamma, calcium channel, voltage-dependent, gamma subunit 1, L-type calcium channel gamma polypeptide, neuronal dihydropyridine-sensitive calcium channel gamma subunit, voltage-dependent calcium channel gamma-1 subunit
PBS & 2% Sucrose. with No Preservative
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit