Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNG7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$153.04 - $436.00
Specifications
Antigen | CACNG7 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 10-50 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Description
CACNG7 Polyclonal specifically detects CACNG7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CACNG7 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 10-50 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
calcium channel, voltage-dependent, gamma subunit 7, Neuronal voltage-gated calcium channel gamma-7 subunit, voltage-dependent calcium channel gamma-7 subunit | |
CACNG7 | |
IgG | |
IgG purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
Sterile water with No Preservative | |
59284 | |
A synthetic peptide from the n-terminal region of human CACNG7 conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in mouse and rat. | |
Primary | |
Reconstitute with 0.5 ml sterilized water. | |
Appears to be specific for CACNG7. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title