Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cacodylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
250g
Packaging:
Glass bottle
75-60-5
C2H7AsO2
137.998
MFCD00002095
OGGXGZAMXPVRFZ-UHFFFAOYSA-N
cacodylic acid,hydroxydimethylarsine oxide,chexmate,silvisar,phytar,arsan,dimethylarsenic acid,erase,arsinic acid, dimethyl,silvisar 510
2513
CHEBI:48765
dimethylarsinic acid
C[As](=O)(C)O
75-60-5
C2H7AsO2
137.998
MFCD00002095
OGGXGZAMXPVRFZ-UHFFFAOYSA-N
cacodylic acid,hydroxydimethylarsine oxide,chexmate,silvisar,phytar,arsan,dimethylarsenic acid,erase,arsinic acid, dimethyl,silvisar 510
2513
CHEBI:48765
dimethylarsinic acid
C[As](=O)(C)O
Assay Percent Range 98%
Linear Formula (CH3)2As(O)OH
Merck Index 15, 1609
Formula Weight 138.01
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 195°C to 198°C
Color White
Quantity 5g
Chemical Name or Material Cacodylic acid
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger